Learn More
claudin 1, Mouse, Clone: 1C5-D9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CLDN1.
Supplier: Abnova Corporation H00009076M01
Description
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. These junctions are comprised of sets of continuous networking strands in the outwardly facing cytoplasmic leaflet, with complementary grooves in the inwardly facing extracytoplasmic leaflet. The protein encoded by this gene, a member of the claudin family, is an integral membrane protein and a component of tight junction strands. Loss of function mutations result in neonatal ichthyosis-sclerosing cholangitis syndrome. [provided by RefSeq
Sequence: MANAGLQLLGFILAFLGWIGAIVSTALPQWRIYSYAGDNIVTAQAMYEGLWMSCVSQSTGQIQCKVFDSLLNLSSTLQATRALMVVGILLGVIAIFVATVGMKCMKCLEDDEVQKMRMAVIGGAIFLLAGLAILVATAWYGNRIVQEFYDPMTPVNARYEFGQALFTGWAAASLCLLGGALLCCSCPRKTTSYPTPRPYPKPAPSSGKDYVSpecifications
claudin 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC012471 | |
CLDN1 | |
CLDN1 (AAH12471.1, 1 a.a. ∼ 211 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Cell Adhesion | |
9076 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
1C5-D9 | |
Mouse monoclonal antibody raised against a full length recombinant CLDN1. | |
CLDN1 | |
CLD1/ILVASC/SEMP1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.