Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CLDN1 Clone: 1C5-D9,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ CB1046100UG
Description
Specifically detects CLDN1 Clone: 1C5-D9 in Human samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Immuno Blotting
CLDN1 is a member of the claudin family, is an integral membrane protein and a component of tight junction strands. Loss of function mutations results in neonatal ichthyosis-sclerosing cholangitis syndrome.
Specifications
CLDN1 | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human | |
Purified |
ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin) | |
1C5-D9 | |
In PBS, pH 7.2. | |
Full-length, recombinant, human CLDN1 (aa 1-212), expressed as a GST fusion protein | |
RUO | |
Recognizes the ∽17kDa CLDN1 protein in PRC/PRF/5 and A431 cells and human small intestine and colon tissue. | |
−20°C | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction