Learn More
claudin 10, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human CLDN10 protein.
Supplier: Abnova Corporation H00009071B01
Description
This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MASTASEIIAFMVSISGWVLVSSTLPTDYWKVSTIDGTVITTATYWANLWKACVTDSTGVSNCKDFPSMLALDGYIQACRGLMIAAVSLGFFGSIFALFGMKCTKVGGSDKAKAKIACLAGIVFILSGLCSMTGCSLYANKITTEFFDPLFVEQKYELGAALFIGWAGASLCIIGGVIFCFSISDNNKTPRYTYNGATSVMSSRTKYHGGEDFKTTNPSKQFDKNAYVSpecifications
claudin 10 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human CLDN10 protein. | |
CLDN10 | |
CPETRL3/OSP-L | |
Mouse | |
50 μL | |
Cell Adhesion | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC010920 | |
CLDN10 | |
CLDN10 (AAH10920, 1 a.a. ∼ 228 a.a) full-length human protein. | |
RUO | |
Yes | |
9071 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.