Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clec12A Rat anti-Mouse, Alexa Fluor 647, Clone: 5D3 (also known as 1/06-5D3), BD

Catalog No. BDB564794 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564794 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564794 Supplier BD Biosciences Supplier No. 564794
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 5D3 monoclonal antibody specifically binds to mouse C-type lectin domain family 12 member A (Clec12A), also known as myeloid inhibitory C-type lectin-like receptor (MICL), C-type lectine-like molecule 1 (CLL-1) or Dendritic cell-associated lectin 2 (DCAL-2). Clec12A is expressed on plasmacytoid dendritic cells, subsets of myeloid dendritic cells, monocytes, macrophage and B cells. Clec12A has one single C-type lectin-like domain and one cytoplasmic immunoreceptor tyrosine-based inhibitory motif. Clec12A reportedly serves as a negative regulator of granulocyte and monocyte function.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Clec12A
Applications Flow Cytometry
Classification Monoclonal
Clone 5D3 (also known as 1/06-5D3)
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description CL12A; C-type lectin domain family 12 member A; CLL-1; DCAL-2; KLRL1; MICL
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Clec12A Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.