Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clec12A Mouse anti-Human, FITC, Clone: 50C1, BD

Catalog No. BDB562569 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562569 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562569 Supplier BD Biosciences Supplier No. 562569
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 50C1 monoclonal antibody specifically binds to human Clec12A (C-type lectin domain family 12 member A). Clec12A is also known as myeloid inhibitory C-type lectin-like receptor (MICL), C-type lectin-like molecule 1 (CLL-1) or dendritic cell-associated lectin 2 (DCAL-2). It is expressed on a variety of cells including monocytes, macrophages, dendritic cells, and granulocytes and perhaps some NK cells. Cleck12A is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. It is a 30kDa type II transmembrane glycoprotein that has one single C-type lectin-like domain and one cytoplasmic immunoreceptor tyrosine-based inhibitory motif (ITIM). Clec12A has similarity with the β-glucan receptor (Dectin-1) and LOX-1 with high N-glycosylation. There are at least five isoforms due to alternative transcript splicing. Signaling through Clec12A can induce internalization of Cleck12A, dendritic cell maturation and the production of cytokines including IL-12. Clec12A may also serve as a negative regulator of activated leukocytes recruited to sites of inflammation

Host Species: Mouse
Clone: 50C1
Isotype: IgG2a, κ
Species Reactivity: Human
Immunogen: Human CLEC12A Transfected Cell Line

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Clec12A
Applications Flow Cytometry
Classification Monoclonal
Clone 50C1
Conjugate FITC
Description CD371; Clec12A; MICL; CLL-1; DCAL-2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CLEC12A Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.