Learn More
C-type lectin domain family 1, member B, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CLEC1B protein.
Supplier: Abnova Corporation H00051266D01P
Description
Natural killer (NK) cells express multiple calcium-dependent (C-type) lectin-like receptors, such as CD94 (KLRD1; MIM 602894) and NKG2D (KLRC4; MIM 602893), that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells.[supplied by OMIM
Sequence: MQDEDGYITLNIKTRKPALVSAVMQRNYLQDENENRTGTLQQLAKRFCQYVVKQSELKGTFKGHKCSPCDTNWRYYGDSCYGFFRHNLTWEESKQYCTDMNATLLKIDNRNIVEYIKARTHLIRWVGLSRQKSNEVWKWEDGSVISENMFEFLEDGKGNMNCAYFHNGKMHPTFCENKHYLMCERKAGMTKVDQLPSpecifications
C-type lectin domain family 1, member B | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human CLEC1B protein. | |
CLEC1B | |
1810061I13Rik/CLEC2/CLEC2B/PRO1384/QDED721 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC029554 | |
CLEC1B | |
CLEC1B (AAH29554.1, 1 a.a. ∼ 196 a.a) full-length human protein. | |
100 μg | |
Membrane Receptors | |
51266 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.