Learn More
C-type lectin domain family 2, member D, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CLEC2D protein.
Supplier: Abnova Corporation H00029121D01P
Description
This gene encodes a member of the natural killer cell receptor C-type lectin family. The encoded protein inhibits osteoclast formation and contains a transmembrane domain near the N-terminus as well as the C-type lectin-like extracellular domain. Several alternatively spliced transcript variants have been identified, but the full-length nature of every transcript has not been defined. [provided by RefSeq
Sequence: MHDSNNVEKDITPSELPANPGCLHSKEHSIKATLIWRLFFLIMFLTIIVCGMVAALSAIRANCHQEPSVCLQAACPESWIGFQRKCFYFSDDTKNWTSSQRFCDSQDADLAQVESFQELNFLLRYKGPSDHWIGLSREQGQPWKWINGTEWTRQFPILGAGECAYLNDKGASSARHYTERKWICSKSDIHVSpecifications
C-type lectin domain family 2, member D | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human CLEC2D protein. | |
CLEC2D | |
CLAX/LLT1/OCIL | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_013269 | |
CLEC2D | |
CLEC2D (NP_037401.1, 1 a.a. ∼ 191 a.a) full-length human protein. | |
100 μg | |
Membrane Receptors | |
29121 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.