Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-type lectin domain family 4, member D, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016642 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-642 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-642 Supplier Abnova Corporation Supplier No. H00338339B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CLEC4D protein.

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region. [provided by RefSeq

Sequence: MGLEKPQSKLEGGMHPQLIPSVIAVVFILLLSVCFIASCLVTHHNFSRCKRGTGVHKLEHHAKLKCIKEKSELKSAEGSTWNCCPIDWRAFQSNCYFPLTDNKTWAESERNCSGMGAHLMTISTEAEQNFIIQFLDRRLSYFLGLRDENAKGQWRWVDQTPFNPRRVFWHKNEPDNSQGENCVVLVYNQDKWAWNDVPCNFEASRICKIPGTTLN

Specifications

Antigen C-type lectin domain family 4, member D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CLEC4D protein.
Formulation PBS with no preservative; pH 7.4
Gene CLEC4D
Gene Accession No. NM_080387.4
Gene Alias CLEC-6/CLEC6/CLECSF8/MCL/MGC40078/MPCL
Gene Symbols CLEC4D
Host Species Mouse
Immunogen CLEC4D (NP_525126.2, 1 a.a. ∼ 215 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338339
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.