Learn More
Clec9A Rat anti-Mouse, Alexa Fluor 647, Clone: 10B4, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 564272
Description
The 10B4 monoclonal antibody specifically binds to mouse Clec9A. Mouse Clec9A (C-type lectin domain family member 9A) is also known as DNGR1 (Dendritic cell natural killer lectin group receptor 1). It is a type II membrane protein with a single extracellular C-type lectin domain. Clec9A is a dendritic cell subtype-restricted C-type lectin-like receptor. Clec9A is selectively expressed on plasmacytoid dendritic cells and CD8+ myeloid dendritic cells. Clec9A reportedly serves as a receptor for necrotic cells. It can mediate the cross-presentation of dead-cell associated antigens in a Syk-dependent manner.

Specifications
Clec9A | |
Monoclonal | |
0.2mg/mL | |
CLC9A; Clec9a; DNGR-1; C-type lectin domain family 9 member A | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
10B4 | |
Alexa Fluor 647 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Clec9A Peptide | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.