Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clec9A Rat anti-Mouse, Brilliant Violet 421, Clone: 10B4, BD

Catalog No. BDB564271 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564271 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564271 Supplier BD Biosciences Supplier No. 564271
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 10B4 monoclonal antibody specifically binds to mouse Clec9A. Mouse Clec9A (C-type lectin domain family member 9A) is also known as DNGR1 (Dendritic cell natural killer lectin group receptor 1). It is a type II membrane protein with a single extracellular C-type lectin domain. Clec9A is a dendritic cell subtype-restricted C-type lectin-like receptor. Clec9A is selectively expressed on plasmacytoid dendritic cells and CD8+ myeloid dendritic cells. Clec9A reportedly serves as a receptor for necrotic cells. It can mediate the cross-presentation of dead-cell associated antigens in a Syk-dependent manner.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Clec9A
Applications Flow Cytometry
Classification Monoclonal
Clone 10B4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description CLC9A; Clec9a; DNGR-1; C-type lectin domain family 9 member A
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Clec9A Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.