Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clec9A Rat anti-Human, Brilliant Violet 421, Clone: 3A4/Clec9A, BD

Catalog No. BDB564266 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564266 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564266 Supplier BD Biosciences Supplier No. 564266
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 3A4/Clec9A monoclonal antibody specifically binds to human Clec9A (C-type lectin domain family member 9A). Clec9A is a type II transmembrane glycoprotein with a single extracellular C-type lectin domain. Clec9A is also known as CD370 and DNGR1 (Dendritic cell natural killer lectin group receptor 1). Clec9A is selectively expressed on CD141+ myeloid dendritic cells. Clec9A reportedly functions as an endocytic receptor for necrotic cells. It can mediate the cross-presentation of dead-cell associated antigens by dendritic cells in a Syk-dependent manner. The 3A4 antibody reportedly crossreacts with peripheral blood dendritic cells form Pigtail macaques ( Macaca nemestrina ).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Clec9A
Applications Flow Cytometry
Classification Monoclonal
Clone 3A4/Clec9A
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description CD370; C-type lectin domain family 9 member A; CLEC9A; DNGR1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Host Species Rat
Immunogen Human Clec9A Peptide
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.