Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC9A Rat anti-Mouse, Clone: 10B4, MilliporeSigma™

Catalog No. 04148MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-148-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-148-MI Supplier MilliporeSigma Supplier No. 04148
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect CLEC9A using this rat monoclonal antibody, Anti-CLEC9A Antibody, clone 10B4 validated for use in western blotting and Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC9A
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 10B4
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001192292
Gene Symbols Clec9a, Dngr-1
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to mouse CLEC9A.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.