Learn More
CLIC1 (M01), Mouse anti-Human, Clone: 2D4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CLIC1.
Supplier: Abnova Corporation H00001192M01
Description
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 1 is a member of the p64 family; the protein localizes principally to the cell nucleus and exhibits both nuclear and plasma membrane chloride ion channel activity. [provided by RefSeq
Sequence: MAEEQPQVELFVKAGSDGAKIGNCPFSQRLFMVLWLKGVTFNVTTVDTKRRTETVQKLCPGGQLPFLLYGTEVHTDTNKIEEFLEAVLCPPRYPKLAALNPESNTAGLDIFAKFSAYIKNSSPALNDNLEKGLLKALKVLDNYLTSPLPEEVDETSAEDEGVSQRKFLDGNELTLADCNLLPKLHIVQVVCKKYRGFTIPEAFRGVHRYLSNAYAREEFASTCPDDEEIELAYEQVAKALKSpecifications
CLIC1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC064527 | |
CLIC1 | |
CLIC1 (AAH64527.1, 1 a.a. ∼ 241 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
2D4 | |
Mouse monoclonal antibody raised against a full-length recombinant CLIC1. | |
CLIC1 | |
G6/NCC27 | |
Mouse | |
Affinity chromatography | |
RUO | |
1192 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.