Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLIC2, Mouse, Clone: 2C1, Abnova™

Catalog No. 89021110 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-110 Supplier Abnova Corporation Supplier No. H00001193M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CLIC2.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 2 is a member of the p64 family; the protein is detected in fetal liver and adult skeletal muscle tissue. This gene maps to the candidate region on chromosome X for incontinentia pigmenti. [provided by RefSeq

Sequence: MSGLRPGTQVDPEIELFVKAGSDGESIGNCPFCQRLFMILWLKGVKFNVTTVDMTRKPEELKDLAPGTNPPFLVYNKELKTDFIKIEEFLEQTLAPPRYPHLSPKYKESFDVGCNLFAKFSAYIKNTQKEANKNFEKSLLKEFKRLDDYLNTPLLDEIDPDSAEEPPVSRRLFLDGDQLTLADCSLLPKLNITKVAAKKYRDFDIPAEFSGVWRYLHNAYAREEFTHTCPEDKEIENTYANVAKQKS

Specifications

Antigen CLIC2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant CLIC2.
Formulation PBS with no preservative; pH 7.4
Gene CLIC2
Gene Accession No. BC022305
Gene Alias CLIC2b/XAP121
Gene Symbols CLIC2
Host Species Mouse
Immunogen CLIC2 (AAH22305, 1 a.a. ∼ 247 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1193
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.