Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chloride intracellular channel 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006689 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-689 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-689 Supplier Abnova Corporation Supplier No. H00009022B01

Mouse polyclonal antibody raised against a full-length human CLIC3 protein.

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 3 is a member of the p64 family and is predominantly localized in the nucleus and stimulates chloride ion channel activity. In addition, this protein may participate in cellular growth control, based on its association with ERK7, a member of the MAP kinase family. [provided by RefSeq

Sequence: MAETKLQLFVKASEDGESVGHCPSCQRLFMVLLLKGVPFTLTTVDTRRSPDVLKDFAPGSQLPILLYDSDAKTDTLQIEDFLEETLGPPDFPSLAPRYRESNTAGNDVFHKFSAFIKNPVPAQDEALYQQLLRALARLDSYLRAPLEHELAGEPQLRESRRRFLDGDRLTLADCSLLPKLHIVDTVCAHFRQAPIPAELRGVRRYLDSAMQEKEFKYTCPHSAEILAAYRPAVHPR

Specifications

Antigen chloride intracellular channel 3
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CLIC3 protein.
Formulation No additive
Gene CLIC3
Gene Accession No. NM_004669.2
Gene Symbols CLIC3
Host Species Mouse
Immunogen CLIC3 (NP_004660.2, 1 a.a. ∼ 236 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9022
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.