Learn More
Description
This gene encodes a protein belonging to the serine/threonine type protein kinase family. This protein is a nuclear dual-specificity kinase that regulates the intranuclear distribution of the serine/arginine-rich (SR) family of splicing factors. Two transcript variants encoding different isoforms have been found for this gene. Related pseudogenes are located on chromosomes 1 and 9. [provided by RefSeq
Specifications
Specifications
| Antigen | CLK3 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 7D6 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant CLK3. |
| Formulation | ascites with no preservative |
| Gene | CLK3 |
| Gene Accession No. | BC002555 |
| Gene Alias | FLJ22858/PHCLK3/PHCLK3/152 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.