Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ceroid-lipofuscinosis, neuronal 3, Mouse, Clone: 1G10, Abnova™

Catalog No. 89014077 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-077 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-077 Supplier Abnova Corporation Supplier No. H00001201M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant CLN3.

This gene encodes a protein that is involved in lysosomal function. Mutations in this, as well as other neuronal ceroid-lipofuscinosis (CLN) genes, cause neurodegenerative diseases commonly known as Batten disease or collectively known as neuronal ceroid lipofuscinoses (NCLs). Many alternatively spliced transcript variants have been found for this gene. [provided by RefSeq

Sequence: MGGCAGSRRRFSDSEGEETVPEPRLPLLDHQGAHWKNAVGFWLLGLCNNFSYVVMLSAAHDILSHKRTSGNQSHVDPGPTPIPHNSSSRFDCNSVSTAAVLLADILPTLVIKLLAPLGLHLLPYSPRVLVSGICAAGSFVLVAFSHSVGTSLCGVVFASISSGLGEVTFLSLTAFYPRAVISWWSSGTGGAGLLGALSYLGLTQAGLSPQQTLLSMLGIPALLLASYFLLLTSPEAQDPGGEEEAESAARQPLIRTEAPESKPGSSSSLSLRERWTVFKGLLWYIVPLVVVYFAEYFINQGLFELLFFWNTSLSHAQQYRWYQMLYQAGVFASRSSLRCCRIRFTWALALLQCLNLVFLLADVWFGFLPSIYLVFLIILYEGLLGGAAYVNTFHNIALETSDEHREFAMAATCISDTLGISLSGLLALPLHDFLCQLS

Specifications

Antigen ceroid-lipofuscinosis, neuronal 3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1G10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant CLN3.
Formulation PBS with no preservative; pH 7.4
Gene CLN3
Gene Accession No. BC002394
Gene Alias BATTENIN/BTS/JNCL/MGC102840
Gene Symbols CLN3
Host Species Mouse
Immunogen CLN3 (AAH02394.1, 1 a.a. ∼ 438 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1201
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.