Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

colipase, pancreatic, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005150 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-150 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-150 Supplier Abnova Corporation Supplier No. H00001208B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CLPS protein.

The protein encoded by this gene is a cofactor needed by pancreatic lipase for efficient dietary lipid hydrolysis. It binds to the C-terminal, noncatalytic domain of lipase, thereby stabilizing an active conformation and considerably increasing the overall hydrophobic binding site. The gene product allows lipase to anchor noncovalently to the surface of lipid micelles, counteracting the destabilizing influence of intestinal bile salts. This cofactor is only expressed in pancreatic acinar cells, suggesting regulation of expression by tissue-specific elements. [provided by RefSeq

Sequence: MEKILILLLVALSVAYAAPGPRGIIINLENGELCMNSAQCKSNCCQHSSALGLARCTSMASENSECSVKTLYGIYYKCPCERGLTCEGDKTIVGSITNTNFGICHDAGRSKQ

Specifications

Antigen colipase, pancreatic
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CLPS protein.
Formulation No additive
Gene CLPS
Gene Accession No. NM_001832.2
Gene Symbols CLPS
Host Species Mouse
Immunogen CLPS (NP_001823.1, 1 a.a. ∼ 112 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 1208
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.