Learn More
CMTM6, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human CMTM6 peptide using ARM Technology.
Supplier: Abnova Corporation H00054918K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
CMTM6 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
CMTM6 | |
CMTM6 | |
A synthetic peptide of human CMTM6 | |
RUO | |
54918 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
CKLF-like MARVEL transmembrane domain containing 6 | |
CMTM6 | |
CKLFSF6, FLJ20396, PRO2219 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.