Learn More
p21, clone: F155-227, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA191045
Description
This gene encodes a potent cyclin-dependent kinase inhibitor. The encoded protein binds to and inhibits the activity of cyclin-CDK2 or -CDK4 complexes, and thus functions as a regulator of cell cycle progression at G1. The expression of this gene is tightly controlled by the tumor suppressor protein p53, through which this protein mediates the p53-dependent cell cycle G1 phase arrest in response to a variety of stress stimuli. This protein can interact with proliferating cell nuclear antigen (PCNA), a DNA polymerase accessory factor, and plays a regulatory role in S phase DNA replication and DNA damage repair. This protein was reported to be specifically cleaved by CASP3-like caspases, which thus leads to a dramatic activation of CDK2, and may be instrumental in the execution of apoptosis following caspase activation. Multiple alternatively spliced variants have been found for this gene.
MA1-91045 detects cN RAS from rat, mouse, human samples.MA1-91045 has been successfully used in immunohistochemistry (frozen tissue), immunohistochemistry (paraffin tissue), Western blot, immunoprecipitation applications.The MA1-91045 immunogen is: Recombinant p21 protein.
Specifications
| cN RAS | |
| Monoclonal | |
| 0.1mg/mL | |
| CDKN1A | |
| CAP20, CDKN1, CDNK1A, MDA-6, SDI1, WAF1, p21CIP1, CDK-interacting protein 1, CDK-interaction protein 1, DNA synthesis inhibitor, cyclin-dependent kinase inhibitor 1, melanoma differentiation associated protein 6, wild-type p53-activated fragment 1 | |
| Mouse | |
| Purified | |
| RUO | |
| 1026 | |
| Store at 4°C, do not freeze | |
| Purified |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| F155-227 | |
| Unconjugated | |
| P38936, P39689 | |
| CDKN1A | |
| Recombinant p21 protein. | |
| 25 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.