Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNDP dipeptidase 2 (metallopeptidase M20 family), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018524 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-524 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-524 Supplier Abnova Corporation Supplier No. H00055748D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CNDP2 protein.

CNDP2, also known as tissue carnosinase and peptidase A (EC 3.4.13.18), is a nonspecific dipeptidase rather than a selective carnosinase (Teufel et al., 2003 [PubMed 12473676]).[supplied by OMIM

Sequence: MAALTTLFKYIDENQDRYIKKLAKWVAIQSVSAWPEKRGEIRRMMEVAAADVKQLGGSVELVDIGKQKLPDGSEIPLPPILLGRLGSDPQKKTVCIYGHLDVQPAALEDGWDSEPFTLVERDGKLYGRGSTDDKGPVAGWINALEAYQKTGQEIPVNVRFCLEGMEESGSEGLDELIFARKDTFFKDVDYVCISDNYWLGKKKPCITYGLRGICYFFIEVECSNKDLHSGVYGGSVHEAMTDLILLMGSLVDKRGNILIPGINEAVAAVTEEEHKLYDDIDFDIEEFAKDVGAQILLHSHKKDILMHRWRYPSLSLHGIEGAFSGSGAKTVIPRKVVGKFSIRLVPNMTPEVVGEQVTSYLTKKFAELRSPNEFKVYMGHGGKPWVSDFSHPHYLAGRRAMKTVFGVEPDLTREGGSIPVTLTFQEATGKNVMLLPVGSADDGAHSQNEKLNRYNYIEGTKMLAAYLYEVSQLKD

Specifications

Antigen CNDP dipeptidase 2 (metallopeptidase M20 family)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CNDP2 protein.
Formulation PBS with no preservative; pH 7.4
Gene CNDP2
Gene Accession No. BC001375
Gene Alias CN2/CPGL/FLJ10830/HsT2298/PEPA
Gene Symbols CNDP2
Host Species Rabbit
Immunogen CNDP2 (AAH01375.1, 1 a.a. ∼ 475 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55748
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.