Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNGA3 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1561450UL
Description
Anti-CNGA3 Antibody detects level of CNGA3 and has been published and validated for use in immunohistochemistry and western blotting.
Specifications
CNGA3 | |
Polyclonal | |
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min). | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat |
Immunohistochemistry, Western Blot | |
Unconjugated | |
CNGA3; CNCG3; CCNCalpha; CNG3; CCNCa; ACHM2; CCNC1; CNG-3 | |
Purified peptide from the C-terminus of rat CNGA3 (Accession number Q9QWN7). | |
50 μL | |
NM_001079878.1; NM_001298.2 | |
Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction