Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPase Mouse anti-Bovine, Canine, Human, Mouse, Pig, Rat, Rabbit, Sheep, Clone: 11-5B, MilliporeSigma™

Catalog No. MAB326MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB326MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB326MI Supplier MilliporeSigma Supplier No. MAB326
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect CNPase using this Anti-CNPase Antibody, clone 11-5B validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNPase
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 11-5B
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.02M phosphate buffer, 0.25MNaCl with 0.1% sodium azide
Gene Accession No. NM_033133.4
Gene Symbols CNP, CNPase, CNP1, EC 3.1.4.37
Host Species Mouse
Immunogen Purified human brain CNPase
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Mouse, Pig, Rabbit, Rat, Sheep
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.