Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

contactin associated protein-like 4, Mouse, Clone: 5G1, Abnova™

Catalog No. 89001558 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-558 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-558 Supplier Abnova Corporation Supplier No. H00085445M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CNTNAP4.

This gene product belongs to the neurexin family, members of which function in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, and thrombospondin N-terminal-like domains. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq

Sequence: VLGRILEHSDVDQETALAGAQGFTGCLSAVQLSHVAPLKAALHPSHPDPVTVTGHVTESSCMAQPGTDATSRERTHSFADHSGTIDDREPLANAIKSDSA

Specifications

Antigen contactin associated protein-like 4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5G1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CNTNAP4.
Formulation ascites with no preservative
Gene CNTNAP4
Gene Accession No. NM_033401
Gene Alias CASPR4/KIAA1763
Gene Symbols CNTNAP4
Host Species Mouse
Immunogen CNTNAP4 (NP_207837, 1145 a.a. ∼ 1244 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 85445
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.