Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Co-Rest/RCOR1 Mouse anti-Human, Mouse, Rat, Clone: K72/8, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN486
Description
Anti-Co-Rest/RCOR1 Antibody, clone K72/8 is a highly specific mouse monoclonal antibody, that targets CoRest and has been tested in western blotting, immunohistochemistry and immunocytochemistry.
Specifications
Co-Rest/RCOR1 | |
Monoclonal | |
Unconjugated | |
NP_055971 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
K72/8 | |
Purified mouse monoclonal IgG1 in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
RCOR1, KIAA0071, RCOR | |
Recombinant protein corresponding to human Co-Rest/RCOR1. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction