Learn More
COASY, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human COASY peptide using ARM Technology.
Supplier: Abnova Corporation H00080347K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. COASY is a bifunctional enzyme that catalyzes the 2 last steps in CoA synthesis. These activities are performed by 2 separate enzymes, phosphopantetheine adenylyltransferase (PPAT; EC 2.7.7.3) and dephospho-CoA kinase (DPCK; EC 2.7.1.24), in prokaryotes (Daugherty et al., 2002 [PubMed 11923312]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
COASY | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
COASY | |
COASY | |
A synthetic peptide of human COASY | |
RUO | |
80347 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
Coenzyme A synthase | |
COASY | |
DPCK, FLJ35179, NBP, PPAT, UKR1, pOV-2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.