Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COBL Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523906
Description
This antibody is predicted to react with rat based on sequence homology.

Specifications
COBL | |
Polyclonal | |
COBL | |
protein cordon-bleu | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12808, 23242 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
O75128, Q5NBX1 | |
COBL | |
KLH conjugated synthetic peptide between 184-211 amino acids from the N-terminal region of human COBL | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction