Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cofilin Mouse, Unlabeled, Clone: 32, BD

Catalog No. BDB612145 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612145 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612145 Supplier BD Biosciences Supplier No. 612145
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cell motility is a basic cellular behavior involved in embryogenesis, neurite outgrowth, wound healing, inflammation, and cancer metastasis. Filamentous actin is an essential component of cell protrusions involved in cell motility. The protrusion formation is regulated by actin polymerization and depolymerization. Cofilin is a ubiquitously expressed G- and F-actin binding protein that contains a nuclear localization signal, a C-terminal hexapeptide sequence that is identical to tropomyosin, and other regions that are homologous to actin binding proteins. In vitro, cofilin has actin severing activity and induces an increased off-rate from the pointed end of actin filaments. These activities can increase the rate of actin polymerization and depolymerization, as well as increase the number of barbed ends available for polymerization. Stimulation of MTLn3 cells with EGF leads to an increase in cofilin at the leading edge of laemellipodia, which correlates with an increase in the number of barbed ends at the leading edge. Cofilin function blocking antibodies inhibit both the appearance of these barbed ends and lamellipodial protrusion. Thus, cofilin is an actin binding protein that regulates actin filament formation during cell motility.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cofilin
Applications Western Blot
Classification Monoclonal
Clone 32
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Cofilin aa. 3-98
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.