Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

component of oligomeric golgi complex 2 (A01), Mouse anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89004546 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-546 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-546 Supplier Abnova Corporation Supplier No. H00022796A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant COG2.

This gene encodes a subunit of the conserved oligomeric Golgi complex that is required for maintaining normal structure and activity of the Golgi complex. The encoded protein specifically interacts with the USO1 vesicle docking protein and may be necessary for normal Golgi ribbon formation and trafficking of Golgi enzymes. Mutations of this gene are associated with abnormal glycosylation within the Golgi apparatus. Alternative splicing results in multiple transcript variants

Sequence: LSESTHKYYETVSDVLNSVKKMEESLKRLKQARKTTPANPVGPSGGMSDDDKIRLQLALDVEYLGEQIQKLGLQASDIKSFSALAELVAAAKDQATAEQP

Specifications

Antigen component of oligomeric golgi complex 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant COG2.
Formulation 50% glycerol
Gene COG2
Gene Accession No. NM_007357
Gene Alias LDLC
Gene Symbols COG2
Host Species Mouse
Immunogen COG2 (NP_031383, 639 a.a. ∼ 738 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 22796
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.