Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL25A1, Mouse, Clone: 1D2, Abnova™

Catalog No. 89002658 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-658 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-658 Supplier Abnova Corporation Supplier No. H00084570M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant COL25A1.

COL25A1 is a brain-specific membrane-bound collagen. Proteolytic processing releases CLAC, a soluble form of COL25A1 containing the extracellular collagen domains that associates with senile plaques in Alzheimer disease (AD; MIM 104300) brains (Osada et al., 2005 [PubMed 15615705]).[supplied by OMIM

Sequence: NDLQARIAALESAKGAPSIHLLPDTLDHLKTMVQEKVERLLAQKSYEHMAKIRIAREAPSECNCPA

Specifications

Antigen COL25A1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant COL25A1.
Formulation PBS with no preservative; pH 7.4
Gene COL25A1
Gene Accession No. NM_198721
Gene Alias CLAC/CLACP
Gene Symbols COL25A1
Host Species Mouse
Immunogen COL25A1 (NP_942014.1, 57 a.a. ∼ 122 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84570
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.