Learn More
COL2A1, Mouse, Clone: 1E1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant COL2A1.
Supplier: Abnova Corporation H00001280M06
Description
This gene encodes the alpha-1 chain of type II collagen, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia, Stickler syndrome type I, and spondyloepimetaphyseal dysplasia Strudwick type. In addition, defects in processing chondrocalcin, a calcium binding protein that is the C-propeptide of this collagen molecule, are also associated with chondrodysplasia. There are two transcripts identified for this gene. [provided by RefSeq
Sequence: MSAFAGLGPREKGPDPLQYMRADQAAGGLRQHDAEVDATLKSLNNQIESIRSPEGSRKNPARTCRDLKLCHPEWKSGDYWIDPNQGCTLDAMKVFCNMETGETCVYPNPANVPKKNWWSSKSKEKKHIWFGETINGGFHFSYGDDNLAPNTANVQMTFLRLLSTEGSQNITYHCKNSIAYLDEAAGNLKKALLIQGSNDVEIRAEGNSRFTYTALKDGCTKHTGKWGKTVIEYRSQKTSRLPIIDIAPMDIGGPEQEFGVDIGPVCFLSpecifications
COL2A1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC007252 | |
COL2A1 | |
COL2A1 (AAH07252, 1 a.a. ∼ 268 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
1E1 | |
Mouse monoclonal antibody raised against a full-length recombinant COL2A1. | |
COL2A1 | |
ANFH/AOM/COL11A3/MGC131516/SEDC | |
Mouse | |
Affinity chromatography | |
RUO | |
1280 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.