Learn More
COL4A3BP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528797
Description
This gene encodes a kinase that specifically phosphorylates the N-terminal region of the non-collagenous domain of the alpha 3 chain of type IV collagen, known as the Goodpasture antigen. Goodpasture disease is the result of an autoimmune response directed at this antigen. One isoform of this protein is also involved in ceramide intracellular transport. Three transcript variants encoding different isoforms have been found for this gene.
PA5-28797 targets COL4A3BP in IF and WB applications and shows reactivity with Human and mouse samples.The PA5-28797 immunogen is synthetic peptide corresponding to a region within amino acids 561 and 624 of Human COL4A3BP.
Specifications
COL4A3BP | |
Polyclonal | |
Unconjugated | |
Q9Y5P4, Q9EQG9 | |
COL4A3BP | |
Synthetic peptide corresponding to a region within amino acids 561 and 624 of Human COL4A3BP | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1mg/mL | |
COL4A3BP | |
CERT, CERTL, GPBP, STARD11, StAR-related lipid transfer (START) domain containing 11, ceramide transfer protein, ceramide transporter, collagen type IV alpha-3-binding protein, hCERT, lipid-transfer protein CERTL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10087, 68018 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.