Learn More
Collagen Type I Goat anti-Bovine, Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Collagen Type I |
| Applications | Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity Purified immunoglobulin. Prior to purification the antisera was adsorbed against collagen types II, III, IV, V and VI immobilized on Sepharose™ 4B.. Liquid in 0.5 mL of 100mM borate buffered saline, pH 8.0, no preservatives. |
| Gene Symbols | COL1A1; OI4 |
| Host Species | Goat |
| Immunogen | Human placental collagen type I isolated by limited pepsin digestion and selective salt precipitation. |
| Purification Method | Affinity Purified |
| Quantity | 200 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.