Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Type I Collagen Goat anti-Human, Mouse, Rat, Bovine, Canine, Chicken, Chinchilla, Elephant, Feline, Guinea Pig, Hamster, Porcine, Rabbit, Sheep, Tree Shrew, Biotin, Polyclonal, Southern Biotech™
SDP

Catalog No. OB131008 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1310-08 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1310-08 Supplier Southern Biotechnology Associates Supplier No. 131008
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SouthernBiotech is a leading provider of antibodies and proteins for extracellular matrix research.

Specifications

Antigen Type I Collagen
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.4mg/mL
Conjugate Biotin
Formulation Supplied in PBS/NaN3
Host Species Goat
Immunogen Type I collagen
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Chinchilla, Elephant, Feline, Guinea Pig, Hamster, Human, Mouse, Pig, Rabbit, Rat, Sheep, Shrew
Content And Storage Store at 2-8°C
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.