Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type II Mouse anti-Chicken, Human, Mouse, Salamander, Clone: 6B3, MilliporeSigma™

Catalog No. MAB8887MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8887MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8887MI Supplier MilliporeSigma Supplier No. MAB8887
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Collagen Type II Antibody, clone 6B3 is an antibody against Collagen Type II for use in immunofluoresence, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen Type II
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6B3
Conjugate Unconjugated
Formulation Purified immunoglobulin in 10mM PBS, pH 7.4 with 0.2% BSA and 0.09% sodium azide.
Gene Accession No. NM_001844.4; NM_033150.2
Gene Symbols COL2A1, SEDC, COL11A3, ANFH, MGC131516, chondrocalcin, AOM
Host Species Mouse
Immunogen Purified type II collagen; obtained from adult chicken sternum by limited pepsinization.
Purification Method Protein G purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Salamander
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.