Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type II Mouse anti-Bovine, Canine, Human, Mouse, Pig, Rat, Sheep, Clone: COLL-II, MilliporeSigma™

Catalog No. MAB1330MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1330MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1330MI Supplier MilliporeSigma Supplier No. MAB1330

Mouse Monoclonal Antibody

This Anti-Collagen Type II Antibody, clone COLL-II is validated for use in ELISA, immunohistochemistry for the detection of Collagen Type II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen Type II
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone COLL-II
Conjugate Unconjugated
Formulation Ascites fluid containing no preservatives.
Gene Accession No. NM_001844.4; NM_033150.2
Gene Symbols COL2A1, SEDC, COL11A3, ANFH, MGC131516, chondrocalcin, AOM
Host Species Mouse
Immunogen Human (cartilage specific) CNBr-cleaved collagen II.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Mouse, Pig, Rat, Sheep
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.