Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type III Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB747MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB747MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB747MI Supplier MilliporeSigma Supplier No. AB747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Collagen Type III Antibody is validated for use in ELISA, immunofluorescence for the detection of Collagen Type III.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen Type III
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation 500ul containing of 0.15M NaCl, 10mM sodium phosphate pH 7.5, with 0.1% mannitol and o.1% dextran as stabilizers contains no preservatives.

Immunoglobulin fraction was prepared by ammonium sulfate precipitation and chromatography on DEAE-cellulose, of
Gene Symbols COL3A1; EDS4A; FLJ34534
Host Species Rabbit
Immunogen Human placental collagen type III
Quantity 500 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000090.3
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.