Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type V Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB763PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB763PMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB763PMI Supplier MilliporeSigma Supplier No. AB763P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Collagen Type V Antibody detects level of Collagen Type V and has been published and validated for use in ELISA, immunofluoresence, RIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen Type V
Applications ELISA, Immunofluorescence, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation IgG fraction. Liquid
Gene Symbols COL5A1; OTTHUMP00000064637
Host Species Rabbit
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000093.3
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.