Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type VII Mouse anti-Human, Clone: LH7.2, MilliporeSigma™

Catalog No. MAB1345MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1345MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1345MI Supplier MilliporeSigma Supplier No. MAB1345
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Collagen Type VII Antibody, C-terminus, clone LH7.2 is an antibody against Collagen Type VII for use in immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen Type VII
Applications Immunocytochemistry
Classification Monoclonal
Clone LH7.2
Conjugate Unconjugated
Formulation Ascites. Liquid with 0.1% sodium azide.
Gene Accession No. NM_000094.3
Gene Symbols COL7A1, EBR1, EBD1, EBDCT
Host Species Mouse
Immunogen Collagen type VII.
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.