Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collapsin Response Mediated Protein 5 Rat anti-Bovine, Human, Rat, Clone: CR-1, MilliporeSigma™

Catalog No. MAB5442MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5442MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5442MI Supplier MilliporeSigma Supplier No. MAB5442
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Collapsin Response Mediated Protein 5 Antibody, clone CR-1 detects level of Collapsin Response Mediated Protein 5 and has been published and validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collapsin Response Mediated Protein 5
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone CR-1
Conjugate Unconjugated
Formulation Unpurified tissue culture supernatant from a perfusion system, filtered through a 0.2μ micron membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL.
Gene Accession No. NM_020134.2
Gene Symbols DPYSL5, CRAM, CRMP5, ULIP6, Ulip6, FLJ45383, DRP-5, CRMP-5
Host Species Rat
Immunogen CRMP-5 purified from rat and human brain.
Quantity 500 μL
Primary or Secondary Primary
Target Species Bovine, Human, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.