Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collapsin Response Mediator Protein 2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9218MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9218MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9218MI Supplier MilliporeSigma Supplier No. AB9218

Rabbit Polyclonal Antibody

Anti-Collapsin Response Mediator Protein 2 Antibody detects level of Collapsin Response Mediator Protein 2 and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collapsin Response Mediator Protein 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid containing 0.1% sodium azide.
Gene Symbols DPYSL2; DHPRP2; CRMP-2; DRP-2; N2A3; CRMP2; DRP2
Host Species Rabbit
Immunogen Synthetic peptide from rat CRMP-2.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001386.4
Target Species Rat
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.