Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
copper metabolism (Murr1) domain containing 1 (A01), Mouse anti-Human, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant COMMD1.
Supplier: Abnova Corporation H00150684A01
Description
COMMD1 is a regulator of copper homeostasis, sodium uptake, and NF-kappa-B (see MIM 164011) signaling (de Bie et al., 2005 [PubMed 16267171]).[supplied by OMIM
Sequence: MAAGELEGGKPLSGLLNALAQDTFHGYPGITEELLRSQLYPEVPPEEFRPFLAKMRGILKSIASADMDFNQLEAFLTAQTKKQGGITSDQAAVISKFWKSHKTKIRESLMNQSRWNSGLRGLSWRVDGKSQSRHSAQIHTPVAIIELELGKYGQESEFLCLEFDEVKVNQILKTLSEVEESISTLISQPNSpecifications
copper metabolism (Murr1) domain containing 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant COMMD1. | |
COMMD1 | |
C2orf5/MGC27155/MURR1 | |
Mouse | |
50 μL | |
Yes | |
150684 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC022046 | |
COMMD1 | |
COMMD1 (AAH22046.1, 1 a.a. ∼ 190 a.a) full-length recombinant protein with GST tag. | |
RUO | |
Primary | |
Human, Mouse | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction