Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Complement 5a, Polyclonal

Catalog No. 2048891ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
20-488-91ML 1 mL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-488-91ML Supplier MilliporeSigma™ Supplier No. 2048891ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Complement 5a Clone: in Human samples, and it is validated for ELISA, Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement 5a
Applications ELISA, Immunoblot, Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation Undiluted serum.
Host Species Rabbit
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Monospecific for human C5a complement component as determined by OT.
Target Species Human
Content And Storage −20°C
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.