Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Complement 5b-9, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 2049031MG
Description
Specifically detects Complement 5b-9 Clone: in Human, Mouse samples, and it is validated for Immunodiffusion, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
C5b-9 is the cytolytically active glycoprotein complex, comprised of C5b, C6, C7, C8 and C9 complement proteins, formed as a result of complement activation via either the classical or alternative pathways.
Specifications
Complement 5b-9 | |
Polyclonal | |
In PBS, pH 7.2. | |
purified human SC5b-9 complex | |
RUO | |
Recognizes complement component complex Cb5-C9. | |
−20°C | |
IgG |
Immunodiffusion, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
1 mg | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction