Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3 Chicken anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB3421MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3421MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3421MI Supplier MilliporeSigma Supplier No. AB3421
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Detect Complement C3 using this Anti-Complement C3 Antibody validated for use in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation Chicken IgY fraction, in 0.15M NaCl, 0.02M NaH2PO4, 0.1% NaN3, pH 7.2.
Gene Symbols C3; ASP; ARMD9; CPAMD1
Host Species Chicken
Immunogen Human C3, full length plasma protein.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000064.2
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.