Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement C3a Mouse anti-Guinea Pig, Human, Rabbit, Clone: H13, MilliporeSigma™

Catalog No. CBL191MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL191MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL191MI Supplier MilliporeSigma Supplier No. CBL191
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Complement C3a Antibody, clone H13 is validated for use in ELISA, immunohistochemistry, western blotting for the detection of Complement C3a.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3a
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone H13
Conjugate Unconjugated
Formulation Protein A purified immunoglobulin lyophilized from PBS, pH 7.4, containing 0.5% BSA and 0.09% sodium azide. Reconstitute with 1 mL distilled water.
Gene Accession No. NM_000064.2
Gene Symbols C3, ASP, ARMD9, CPAMD1
Host Species Mouse
Immunogen Human complement component C3
Quantity 50 μg
Primary or Secondary Primary
Target Species Guinea Pig, Human, Rabbit
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.