Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Complement Component C3aR APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB5995A
This item is not returnable.
View return policy
Description
Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry.Specifications
Complement Component C3aR | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
719 | |
Human | |
Purified |
Flow Cytometry | |
534625 | |
Flow Cytometry 10 uL/10^6 cells | |
Q16581 | |
C3AR1 | |
NS0 mouse myeloma cell line transfected with human C3a R Met1-Val482 Accession # Q16581 | |
100 Tests | |
Primary | |
Detects human Complement Component C3a R in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction