Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Complement Factor H-related 4/CFHR4, Alexa Fluor 488, Clone: 640212, R&D Systems™

Catalog No. IC5980G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5980G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5980G Supplier R&D Systems Supplier No. IC5980G
This item is not returnable. View return policy

Applications:

Flow Cytometry (Intracellular Staining)

Specifications

Antigen Complement Factor H-related 4/CFHR4
Applications Flow Cytometry
Classification Monoclonal
Clone 640212
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q92496
Gene Alias complement factor H-related protein 4, CFHL4, CFHR4, complement factor H-related 4, Complement Factor Hrelated 4, FHR4, FHR-4
Gene Symbols CFHR4
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Complement Factor H-related 4/CFHR4, Glu20-Glu331, Accession # Q92496
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10877
Test Specificity Detects human Complement Factor H-related 4/CFHR4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) CFHR1, rhCFHR2, or rhCFHR5 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.