Learn More
COMT Mouse, Unlabeled, Clone: 4, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 611970
Description
Catechol-O-methyltransferase (COMT) and monoamine oxidase (MAO) are the major mammalian enzymes involved in the degradation of the catecholamine neurotransmitters, dopamine, norepinephrine, and epinephrine. COMT is a Mg2+-dependent enzyme that catalyzes the transfer of methyl groups from S-adenosyl methionine to a hydroxyl group of a catecholic substrate. Two forms of COMT are found in rat brain, a 24kDa soluble COMT (S-COMT) and a 28kDa membrane-bound COMT (MB-COMT). COMT is widely expressed in brain, but its importance in catecholamine neurotransmitter degradation relative to MAO varies in different brain regions. In addition, COMT may function primarily in extraneuronal areas, such as in glial cells and postsynaptic neurons. COMT-deficient mice have sex- and region-specific alterations in dopamine levels in the brain, and display impaired emotional reactivity and aggressive behavior. Thus, COMT-mediated degradation of catecholamines in the brain may have important roles in maintaining normal catecholamine levels, as well as normal social behavior.
Host Species: Mouse
Clone: 4
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Mouse COMT aa. 26-141
Western Blotting

Specifications
COMT | |
Monoclonal | |
250μg/mL | |
Catechol-O-Methyltransferase | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
4 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse COMT aa. 26-141 | |
50 μg | |
Cell Biology | |
Mouse, Rat | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.