Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cone Arrestin Rabbit anti-Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB15282MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15282MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15282MI Supplier MilliporeSigma Supplier No. AB15282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Cone Arrestin, Cat. No. AB15282, is a highly specific rabbit polyclonal antibody that targets Arrestin-C and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cone Arrestin
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in PBS with 0.05% NaN3
Host Species Rabbit
Immunogen Synthetic Linear Peptide
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for 12 months in undiluted aliquotes
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.